Skip to Content
Merck
All Photos(1)

Key Documents

706728

Sigma-Aldrich

4-Allyltoluene

97%

Synonym(s):

1-Allyl-4-methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.509

density

0.865 g/mL at 25 °C

SMILES string

Cc1ccc(CC=C)cc1

InChI

1S/C10H12/c1-3-4-10-7-5-9(2)6-8-10/h3,5-8H,1,4H2,2H3

InChI key

WAEOXIOXMKNFLQ-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.9 °F

Flash Point(C)

54.4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

T Ishida et al.
Enantiomer, 3(2), 133-147 (1998-10-23)
Thirteen kinds of aromatic compounds (Alkyl- and alkenyl-) were administered to rabbits and the chiralities of their optical active metabolite were determined by the synthesis of the enantiomers or their derivatives. This work has revealed many examples of enantioselective biotransformations
T Matsumoto et al.
Biological & pharmaceutical bulletin, 17(11), 1441-1445 (1994-11-01)
4-Isopropenyltoluene (8) was administered orally to rabbits and the following four optically active metabolites, 2-(p-tolyl)propanoic acid (10), p-(1-carboxyethyl)benzoic acid (11), 2-hydroxy-2-(p-tolyl)propanoic acid (12), and 2-(p-tolyl)-1,2-propanediol (13) were isolated from urine in addition to an optically inactive metabolite, 4-isopropenylbenzoic acid (9).
Nurhayat Tabanca et al.
Molecules (Basel, Switzerland), 24(13) (2019-07-03)
Ceratitis capitata, the Mediterranean fruit fly, is one of the most serious agricultural pests worldwide responsible for significant reduction in fruit and vegetable yields. Eradication is expensive and often not feasible. Current control methods include the application of conventional insecticides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service