Skip to Content
Merck
All Photos(1)

Documents

655856

Sigma-Aldrich

4,4,5,5-Tetramethyl-1,3,2-dioxaborolane

97%

Synonym(s):

HBpin, Pinacolborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13BO2
CAS Number:
Molecular Weight:
127.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.396 (lit.)

bp

42-43 °C/50 mmHg (lit.)

density

0.882 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)OBOC1(C)C

InChI

1S/C6H13BO2/c1-5(2)6(3,4)9-7-8-5/h7H,1-4H3

InChI key

UCFSYHMCKWNKAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4,4,5,5-Tetramethyl-1,3,2-dioxaborolane may be used for:
  • Borylation at the benzylic C-H bond of alkylbenzenes in the presence of a palladium catalyst to form pinacol benzyl boronate.
  • Hydroboration of alkyl or aryl alkynes and alkenes in the presence of transition metal catalysts.
  • Coupling with aryl iodides in the presence of a copper catalyst to form aryl boronates.
  • Asymmetric hydroboration of 1,3-enynes to form chiral allenyl boronates.

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed benzylic C?H borylation of alkylbenzenes with bis (pinacolato) diboron or pinacolborane.
Ishiyama T
Chemistry Letters (Jpn), 30(11), 1082-1083 (2001)
A study of hydroboration of alkenes and alkynes with pinacolborane catalyzed by transition metals.
Pereira S
Tetrahedron Letters, 37(19), 3283-3286 (1996)
Formation of arylboronates by a CuI-catalyzed coupling reaction of pinacolborane with aryl iodides at room temperature.
Zhu W
Organic Letters, 8(2), 261-263 (2006)
Copper-Catalyzed Asymmetric Hydroboration of 1, 3-Enynes with Pinacolborane to Access Chiral Allenylboronates.
Sang HL
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry (2018)

Articles

Arylboronic acids and esters, vital tools in chemical transformations, find extensive use, particularly in the Suzuki-Miyaura cross-coupling reaction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service