Skip to Content
Merck
All Photos(1)

Key Documents

693316

Sigma-Aldrich

(S)-2-[[3,5-Bis(trifluoromethyl)phenyl]thioureido]-N-benzyl-N,3,3-trimethylbutanamide

97%

Synonym(s):

(2S)-2-[[[[3,5-Bis(trifluoromethyl)phenyl]amino]thioxomethyl]amino]-N-3,3-trimethyl-N-(phenylmethyl)butanamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$209.00
500 MG
$963.00

$209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$209.00
500 MG
$963.00

About This Item

Empirical Formula (Hill Notation):
C23H25F6N3OS
CAS Number:
Molecular Weight:
505.52
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$209.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

optical activity

[α]20/D -57.0°, c = 1 in chloroform

mp

147-152 °C

functional group

amide
amine
fluoro
phenyl
thiourea

SMILES string

CN(Cc1ccccc1)C(=O)[C@@H](NC(=S)Nc2cc(cc(c2)C(F)(F)F)C(F)(F)F)C(C)(C)C

InChI

1S/C23H25F6N3OS/c1-21(2,3)18(19(33)32(4)13-14-8-6-5-7-9-14)31-20(34)30-17-11-15(22(24,25)26)10-16(12-17)23(27,28)29/h5-12,18H,13H2,1-4H3,(H2,30,31,34)/t18-/m1/s1

InChI key

LTFFYVVCUVYZPE-GOSISDBHSA-N

General description

(S)-2-[[3,5-Bis(trifluoromethyl)phenyl]thioureido]-N-benzyl-N,3,3-trimethylbutanamide is a chiral thiourea organocatalyst developed by the Jacobsen′s group for asymmetric organic synthesis.

Application

Versatile catalyst for reactions including the Mannich reaction.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Divergent Stereoinduction Mechanisms in Urea-Catalyzed Additions to Imines
Wenzel, A.G.; Lalonde, M.P.; Jacobsen, E.N.
Synlett, 1919-1919 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service