Skip to Content
Merck
All Photos(1)

Documents

690384

Sigma-Aldrich

N-[3,5-Bis(trifluoromethyl)phenyl]-N′-[(8a,9S)-10,11-dihydro-6′-methoxy-9-cinchonanyl]thiourea

90%

Synonym(s):

1-[3,5-Bis(trifluoromethyl)phenyl)-3-{(S)[(2S,4S,5R)-5-ethyl-1-aza-bicyclo[2.2.2]oct-2-yl]-(6-methoxy-4-quinolinyl)methyl}thiourea, epi-N-Dihydroquinyl-N′-bis(3,5-trifluoromethyl)phenylthiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H30F6N4OS
CAS Number:
Molecular Weight:
596.63
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥89.0% (HPLC)
90%

form

lumps

optical activity

[α]/D -107.0±5.0°, c = 1 in chloroform

SMILES string

CC[C@H]1CN2CCC1CC2[C@@H](NC(=S)Nc3cc(cc(c3)C(F)(F)F)C(F)(F)F)c4ccnc5ccc(OC)cc45

InChI

1S/C29H30F6N4OS/c1-3-16-15-39-9-7-17(16)10-25(39)26(22-6-8-36-24-5-4-21(40-2)14-23(22)24)38-27(41)37-20-12-18(28(30,31)32)11-19(13-20)29(33,34)35/h4-6,8,11-14,16-17,25-26H,3,7,9-10,15H2,1-2H3,(H2,37,38,41)/t16-,17-,25-,26-/m0/s1

InChI key

KBYUJTLQXUVPQI-FRSFCCSCSA-N

Application

Highly enantioselective conjugate addition of nitromethane to chalcones using bifunctional cinchona organocatalysts

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Urea- and thiourea-substituted cinchona alkaloid derivatives as highly efficient bifunctional organocatalysts for the asymmetric addition of malonate to nitroalkenes: inversion of configuration at C9 dramatically improves catalyst performance.
Séamus H McCooey et al.
Angewandte Chemie (International ed. in English), 44(39), 6367-6370 (2005-09-02)
Benedek Vakulya et al.
Organic letters, 7(10), 1967-1969 (2005-05-07)
Cinchona alkaloid-derived chiral bifunctional thiourea organocatalysts were synthesized and applied in the Michael addition between nitromethane and chalcones with high ee and chemical yields.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service