Skip to Content
Merck
All Photos(1)

Key Documents

691674

Sigma-Aldrich

2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl gold(I) bis(trifluoromethanesulfonyl)imide

Synonym(s):

XPhos AuNTf2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$333.00
1 G
$956.00

$333.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$333.00
1 G
$956.00

About This Item

Empirical Formula (Hill Notation):
C35H49AuF6NO4PS2
CAS Number:
Molecular Weight:
953.83
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$333.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

core: gold
reagent type: catalyst

SMILES string

FC(F)(F)S(=O)(=O)N([Au])S(=O)(=O)C(F)(F)F.CC(C)c1cc(C(C)C)c(c(c1)C(C)C)-c2ccccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C33H49P.C2F6NO4S2.Au/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/h13-14,19-25,27-28H,7-12,15-18H2,1-6H3;;/q;-1;+1

InChI key

GMVGZINKTXAKDT-UHFFFAOYSA-N

Application

Gold Catalysts — 21st Century ′Gold Rush′

  • Catalyst for isomerization of allenyl carbinol esters, hydroxy- and methoxycyclization, cycloisomerisation of diynes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea K Buzas et al.
Organic letters, 9(6), 985-988 (2007-02-14)
A study concerning the gold(I)-catalyzed rearrangement of diversely substituted allenyl carbinol esters into functionalized 1,3-butadien-2-ol esters is described. The mild conditions employed allow the efficient, rapid, and stereoselective synthesis of a variety of such compounds via a new 1,3-shift of
Gold(I)-catalyzed 5-endo hydroxy- and alkoxycyclization of 1,5-enynes: efficient access to functionalized cyclopentenes.
Andrea K Buzas et al.
Angewandte Chemie (International ed. in English), 46(7), 1141-1144 (2006-12-14)
An unusual access to medium sized cycloalkynes by a new gold(I)-catalysed cycloisomerisation of diynes.
Yann Odabachian et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(36), 8966-8970 (2009-08-04)

Articles

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service