Skip to Content
Merck
All Photos(1)

Key Documents

689661

Sigma-Aldrich

(1S,2S)-(+)-Norpseudoephedrine

≥98.0% (NT)

Synonym(s):

(+)-Cathine, (+)-Pseudonorephedrine, (1S,2S)-2-Amino-1-phenyl-1-propanol, D-(+)-Norpseudoephedrine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥98.0% (NT)

form

crystals

optical activity

[α]/D +34.0±2.0°, c = 3.5 in ethanol

drug control

USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

storage temp.

2-8°C

SMILES string

C[C@H](N)[C@@H](O)c1ccccc1

InChI

1S/C9H13NO/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7,9,11H,10H2,1H3/t7-,9+/m0/s1

InChI key

DLNKOYKMWOXYQA-IONNQARKSA-N

Application

(1S,2S)-(+)-Norpseudoephedrine can be used:
  • As a starting material for the synthesis of benzamide based ligands applicable in the Tsuji−Trost allylic alkylation reactions.
  • To prepare its catalyst derivatives, which are applicable in the alkylation of benzaldehydes using diethylzinc.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

β-Hydroxy and β-(o-diphosphino) benzoyloxy (o-diphosphino) benzamides as ligands for asymmetric allylic alkylation
Mahadik GS, et al.
Tetrahedron Asymmetry, 20(10), 1132-1137 (2009)
β-Amino alcohols derived from (1R,2S)-norephedrine and (1S, 2S)-pseudonorephedrine as catalysts in the asymmetric addition of diethylzinc to aldehydes
Parrott II, et al.
Tetrahedron Asymmetry, 19(1), 19-26 (2008)
Kuei Hui Chan et al.
Journal of analytical toxicology, 33(3), 162-166 (2009-04-18)
Sho-seiryu-to is one of the most common Traditional Chinese Medicine preparations for the attenuation of colds. Ephedrae Herba is one of the prescriptions of Sho-seiryu-to. The major ingredients of Ephedrae Herba, ephedrines, are banned substances on the World Anti-Doping Agency
A Pokrywka et al.
International journal of sports medicine, 30(8), 569-572 (2009-04-22)
Pseudoephedrine (PSE) as a sympathomimetic is an ingredient of many proprietary medicines which are available on the medical market over the counter (OTC drugs). It can be converted to cathine (CATH, norpseudoephedrine) inside the body. Until the end of 2003
John S Chappell et al.
Forensic science international, 195(1-3), 108-120 (2009-12-29)
A primary concern with the forensic analysis of the khat plant (Catha edulis) has been the need to preserve the principle psychoactive component, cathinone, which converts to the less-active substance, cathine, after harvesting. The loss of cathinone has serious legal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service