68940
Dichloro(methyl)propylsilane
≥97.0% (GC)
Synonym(s):
Methyl-propyl-dichlorosilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H10Cl2Si
CAS Number:
Molecular Weight:
157.11
Beilstein:
1733841
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥97.0% (GC)
form
liquid
refractive index
n20/D 1.420
density
1.046 g/mL at 20 °C (lit.)
SMILES string
CCC[Si](C)(Cl)Cl
InChI
1S/C4H10Cl2Si/c1-3-4-7(2,5)6/h3-4H2,1-2H3
InChI key
GNVPGBIHGALKRR-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
80.6 °F - closed cup
Flash Point(C)
27 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service