Skip to Content
Merck
All Photos(1)

Key Documents

307297

Sigma-Aldrich

Dichloro(3-chloropropyl)methylsilane

98%

Synonym(s):

(3-Chloropropyl)methyldichlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$129.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$129.00

About This Item

Linear Formula:
Cl(CH2)3Si(CH3)Cl2
CAS Number:
Molecular Weight:
191.56
Beilstein:
1736034
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$129.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

80 °C/18 mmHg (lit.)

density

1.227 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

C[Si](Cl)(Cl)CCCCl

InChI

1S/C4H9Cl3Si/c1-8(6,7)4-2-3-5/h2-4H2,1H3

InChI key

UCJHMXXKIKBHQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Synthetic precursor to enoxysilacyclobutanes.[1]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 116, 7026-7026 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service