Skip to Content
Merck
All Photos(1)

Documents

67705

Sigma-Aldrich

Methyl isocyanate bound to polystyrene HL

extent of labeling: ~1.6 mmol/g isocyanate loading (high loading)

Synonym(s):

Isocyanate polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: amine reactive

extent of labeling

~1.6 mmol/g isocyanate loading (high loading)

matrix

crosslinked with 2% DVB

particle size

200-400 mesh

Other Notes

Scavenger resin to remove excess amines and hydrazines from reaction solutions

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.W. Kaldor et al.
Tetrahedron Letters, 37, 7193-7193 (1996)
R.J. Booth et al.
Journal of the American Chemical Society, 119, 4882-4882 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service