Skip to Content
Merck
All Photos(2)

Key Documents

526118

Sigma-Aldrich

4-(Dimethylamino)benzoyl chloride

97%

Synonym(s):

DMABC

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$61.80
5 G
$137.00

$61.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$61.80
5 G
$137.00

About This Item

Linear Formula:
(CH3)2NC6H4COCl
CAS Number:
Molecular Weight:
183.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

145-149 °C (lit.)

functional group

acyl chloride
amine

SMILES string

CN(C)c1ccc(cc1)C(Cl)=O

InChI

1S/C9H10ClNO/c1-11(2)8-5-3-7(4-6-8)9(10)12/h3-6H,1-2H3

InChI key

UGJDXRVQCYBXAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Dimethylamino)benzoyl chloride is also referred to as 4-(chloroformyl)-N,N-dimethylaniline.
Material is reported to be very moisture sensitive. Handle under an inert atmosphere

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service