Skip to Content
Merck
All Photos(2)

Key Documents

676349

Sigma-Aldrich

3-Phenyl-1H-pyrazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$61.60

$61.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$61.60

About This Item

Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.60


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

78-82 °C

functional group

phenyl

SMILES string

c1ccc(cc1)-c2cc[nH]n2

InChI

1S/C9H8N2/c1-2-4-8(5-3-1)9-6-7-10-11-9/h1-7H,(H,10,11)

InChI key

OEDUIFSDODUDRK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Manzoor et al.
Journal of economic entomology, 110(6), 2527-2533 (2017-10-19)
The aim of this study was to test the bioavailability and degradation in soil of the termiticides bifenthrin and fipronil, which are used to treat subterranean termites (Heterotermes indicola, Wasmann). Soil collected from different areas of Lahore was categorized as
Tianyun Su et al.
Journal of medical entomology, 56(4), 1047-1054 (2019-02-28)
The peridomestic anthropophilic Aedes aegypti L. (Diptera: Culicidae) is originated from the wild zoophilic subspecies Aedes aegypti formosus in sub-Saharan Africa, and currently has a broad distribution in human-modified environments of the tropics and subtropics worldwide. In California, breeding populations
Julie Chevalier et al.
Environmental toxicology and chemistry, 34(8), 1760-1769 (2015-03-12)
Behavior is increasingly reported as a sensitive and early indicator of toxicant stress in aquatic organisms. However, the systematic understanding of behavioral effects and comparisons between effect profiles is hampered because the available studies are limited to few chemicals and
Haihua Wu et al.
Pesticide biochemistry and physiology, 115, 9-14 (2014-10-14)
The acute toxicity of fipronil and its sublethal effects on detoxification enzymes (carboxylesterases (CarEs), glutathione S-transferases (GSTs), and 7-ethoxycoumarin O-deethylase (ECOD)) in zebrafish (Danio rerio) were investigated. The results indicated that the 24-h LC50 of fipronil for zebrafish was 220.4

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service