Skip to Content
Merck
All Photos(1)

Key Documents

673943

Sigma-Aldrich

2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

96%

Synonym(s):

Methoxyboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$58.40
25 G
$183.00

$58.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$58.40
25 G
$183.00

About This Item

Empirical Formula (Hill Notation):
C7H15BO3
CAS Number:
Molecular Weight:
158.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$58.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

refractive index

n20/D 1.4096

density

0.9642 g/mL at 25 °C

SMILES string

COB1OC(C)(C)C(C)(C)O1

InChI

1S/C7H15BO3/c1-6(2)7(3,4)11-8(9-5)10-6/h1-5H3

InChI key

JZZJAWSMSXCSIB-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

108.0 °F - closed cup

Flash Point(C)

42.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service