Skip to Content
Merck
All Photos(1)

Key Documents

672289

Sigma-Aldrich

2,2-Dibenzyl-1,3-propanediol

≥97.0%

Synonym(s):

2,2-Bis(phenylmethyl)-1,3,propanediol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$458.00

$458.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
$458.00

About This Item

Empirical Formula (Hill Notation):
C17H20O2
CAS Number:
Molecular Weight:
256.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$458.00


Estimated to ship on12 April 2025


Request a Bulk Order

Assay

≥97.0% (HPLC)
≥97.0%

mp

84-88 °C

functional group

hydroxyl
phenyl

SMILES string

OCC(CO)(Cc1ccccc1)Cc2ccccc2

InChI

1S/C17H20O2/c18-13-17(14-19,11-15-7-3-1-4-8-15)12-16-9-5-2-6-10-16/h1-10,18-19H,11-14H2

InChI key

AZLRECYOMFCGTK-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service