Skip to Content
Merck
All Photos(1)

Key Documents

554718

Sigma-Aldrich

REV-ERBα Antagonist, SR8278

The REV-ERBα Antagonist, SR8278 controls the biological activity of REV-ERBα.

Synonym(s):

REV-ERBα Antagonist, SR8278, SR8278SR 8278

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$183.00

$183.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$183.00

About This Item

Empirical Formula (Hill Notation):
C18H19NO3S2
CAS Number:
Molecular Weight:
361.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$183.00


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

Assay

>95% (HPLC)

form

gum
solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to yellow-brown

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

n1c2c(ccc1COc3cc(ccc3)C(O)CCCCC)cccc2

InChI key

JRLOEMCOOZSCQP-UHFFFAOYSA-N

General description

A cell-permeable REV-ERB antagonist that inhibits REV-ERBα transcriptional repression activity (EC50 = 0.47 µM) in a REV-ERB target gene assay. It is also shown to block the ability of GSK4112 to enhance REV-ERB-&alpha-dependent repression (EC50 = 0.35 µM) in a co-transfection assay. Treatment of HepG2 cells with SR8278 at 10 µM results in a significant increase in the expression of either G6Pase and PEPCK mRNA expression, consistent with its antagonistic role against endogenous heme.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to months at -20°C.

Other Notes

Kojetin, D., et al. 2010, Chem Bio in press

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang-Yuan Mao et al.
Cellular and molecular gastroenterology and hepatology, 12(2), 395-425 (2021-03-07)
Rev-erbα represents a powerful transcriptional repressor involved in immunity. However, the regulation, function, and clinical relevance of Rev-erbα in Helicobacter pylori infection are presently unknown. Rev-erbα was examined in gastric samples from H pylori-infected patients and mice. Gastric epithelial cells (GECs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service