Skip to Content
Merck
All Photos(1)

Documents

671711

Sigma-Aldrich

(R)-(−)-N-(2,3-Epoxypropyl)phthalimide

≥99.0% (GC)

Synonym(s):

(R)-N-Glycidylphthalimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO3
CAS Number:
Molecular Weight:
203.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

form

solid

optical activity

[α]/D -7 to -13°, c = 2.2 in chloroform

mp

100-102 °C

SMILES string

O=C1N(C[C@@H]2CO2)C(=O)c3ccccc13

InChI

1S/C11H9NO3/c13-10-8-3-1-2-4-9(8)11(14)12(10)5-7-6-15-7/h1-4,7H,5-6H2/t7-/m1/s1

InChI key

DUILGEYLVHGSEE-SSDOTTSWSA-N

Application

(R)-(-)-N-(2,3-Epoxypropyl)phthalimide may be used in the preparation of the following potential chiral solvating agents:
  • 2-[(2R)-2-hydroxy-3-[[(1R)-2-hydroxy-1-phenyl-ethyl]amino]propyl] isoindoline-1,3-dione
  • 2-[(2R)-2-hydroxy-3-[[(1R,2S)-2-hydroxyindan-1-yl]amino]propyl]isoindoline-1,3-dione
  • 2-[(2R)-2-hydroxy-3-[[(1R)-1-(hydroxymethyl)-propyl]amino]propyl] isoindoline-1,3-dione
  • 2-[(2R)-2-hydroxy-3-[[(1S)-1-phenylethyl]-amino]propyl]isoindoline-1,3-dione
It may also be used in the synthesis of 2-((5R)-2-oxo-5-oxazolidinyl) methyl)-1H-isoindole-1,3 (2H)-dione.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile synthesis of 2-((5R)-2-oxo-5-oxazolidinyl) methyl)-1H-isoindole-1, 3 (2H)-dione.
Madhusudhan G, et al.
Indian J. Chem. B, 45, 1264-1268 (2006)
Amino alcohol based chiral solvating agents: synthesis and applications in the NMR enantiodiscrimination of carboxylic acids.
Bozkurt S, et al.
Tetrahedron Asymmetry, 22(5), 541-549 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service