Skip to Content
Merck
All Photos(1)

Key Documents

667668

Sigma-Aldrich

Methyl 1H-benzotriazole-1-carboxylate, 3-oxide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O3
CAS Number:
Molecular Weight:
193.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

128 °C (dec.)

SMILES string

COC(=O)n1n[n+]([O-])c2ccccc12

InChI

1S/C8H7N3O3/c1-14-8(12)10-6-4-2-3-5-7(6)11(13)9-10/h2-5H,1H3

InChI key

FJAORCXXKUNDMQ-UHFFFAOYSA-N

Application

Reactant for:
  • Carbonylation reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service