Skip to Content
Merck
All Photos(1)

Key Documents

667536

Sigma-Aldrich

Methyl 1H-benzotriazole-1-carboxylate

97%

Synonym(s):

Methyl ester 1H-benzotriazole-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

81-84 °C

SMILES string

COC(=O)n1nnc2ccccc12

InChI

1S/C8H7N3O2/c1-13-8(12)11-7-5-3-2-4-6(7)9-10-11/h2-5H,1H3

InChI key

DLEZVOPJBSAAGW-UHFFFAOYSA-N

Application

New Benzotriazole Reagents

  • Reactant for synthesis of azolecarboxylic acid derivatives

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service