Skip to Content
Merck
All Photos(1)

Key Documents

667013

Sigma-Aldrich

trans-3-(Cyclopentyl)-1-propenylboronic acid pinacol ester

97%

Synonym(s):

2-[(1E)-3-Cyclopentyl-1-propen-1-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-(3-Cyclopentylpropen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25BO2
CAS Number:
Molecular Weight:
236.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.456

bp

90-93 °C/0.4 mmHg

density

0.927 g/mL at 25 °C

SMILES string

CC1(C)OB(OC1(C)C)\C=C\CC2CCCC2

InChI

1S/C14H25BO2/c1-13(2)14(3,4)17-15(16-13)11-7-10-12-8-5-6-9-12/h7,11-12H,5-6,8-10H2,1-4H3/b11-7+

InChI key

GXNIAKZVBGUMHU-YRNVUSSQSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service