664774
4-Chlorobenzylsulfonyl chloride
97%
Synonym(s):
(4-Chlorophenyl)methanesulfonyl chloride, 4-Chlorobenzenemethanesulfonyl chloride, p-Chloro-α-toluenesulfonyl chloride
About This Item
Recommended Products
Assay
97%
form
solid
mp
93-97 °C
SMILES string
Clc1ccc(CS(Cl)(=O)=O)cc1
InChI
1S/C7H6Cl2O2S/c8-7-3-1-6(2-4-7)5-12(9,10)11/h1-4H,5H2
InChI key
DBJRPJSDYFDWPV-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Sulfonyl chlorides are often chosen as building blocks in medicinal chemistry for their ability to easily react with heterocyclic amines to create complex sulfonamides.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service