Skip to Content
Merck
All Photos(1)

Key Documents

663905

Sigma-Aldrich

N,N′-Diphenyl-3,4,9,10-perylenedicarboximide

98%

Synonym(s):

PTCDI-Ph

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H18N2O4
CAS Number:
Molecular Weight:
542.54
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

>300 °C

λmax

527 nm

fluorescence

λem ≤534 nm in chloroform

semiconductor properties

N-type (mobility=10−5 cm2/V·s)

SMILES string

O=C1N(c2ccccc2)C(=O)c3ccc4c5ccc6C(=O)N(c7ccccc7)C(=O)c8ccc(c9ccc1c3c49)c5c68

InChI

1S/C36H18N2O4/c39-33-25-15-11-21-23-13-17-27-32-28(36(42)38(35(27)41)20-9-5-2-6-10-20)18-14-24(30(23)32)22-12-16-26(31(25)29(21)22)34(40)37(33)19-7-3-1-4-8-19/h1-18H

InChI key

OGEZSLXPCKHGKO-UHFFFAOYSA-N

General description

N,N′-Diphenyl-3,4,9,10-perylenedicarboximide (PTCDI-Ph) is an asphaltene based conducting polymer that has an archipelago model. It is a perylene diimide derivative that has a high electron affinity and shows good chemical stability. It can be used as an active layer in the development of organic electronic devices.

Application

PTCDI-Ph is a conjugating polymer that forms an ultrathin film that can be used in the fabrication of organic field effect transistors (OFETs) for nitrogen dioxide sensor based applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High performance nitrogen dioxide sensor based on organic field-effect transistor utilizing ultrathin CuPc/PTCDI-C8 heterojunction.
Fan H, et al.
Synthetic Metals, 211(9), 161-166 (2016)
The influence of CO2 on the structure of confined asphaltenes in calcite nanopores.
Mohammed S and Gadikota G
Fuel: The Science and Technology of Fuel and Energy, 236, 769-777 (2019)
Response enhancement mechanism of NO2 gas sensing in ultrathin pentacene field-effect transistors.
Mirza M, et al.
Organic Electronics, 24(9), 96-100 (2015)
Electronic structure of the conduction band of the interface region of ultrathin films of substituted perylenedicarboximides and the germanium oxide surface.
Komolov AS, et al.
Physics of the Solid State, 58(9), 1901-1905 (2016)
Heteroepitaxy growth high performance films of perylene diimide derivatives.
Huang L, et al.
Organic Electronics, 11(2), 195-201 (2010)

Articles

Review the potential of self-assembled multilayer gate dielectric films fabricated from silane precursors for organic, inorganic, and transparent TFT and for TFT circuitry and OLED displays.

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service