Skip to Content
Merck
All Photos(2)

Key Documents

647624

Sigma-Aldrich

4-Amino-2-methoxybenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$102.00

$102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$102.00

About This Item

Linear Formula:
H2NC6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
167.16
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

$102.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

149-153 °C (lit.)

application(s)

peptide synthesis

SMILES string

COc1cc(N)ccc1C(O)=O

InChI

1S/C8H9NO3/c1-12-7-4-5(9)2-3-6(7)8(10)11/h2-4H,9H2,1H3,(H,10,11)

InChI key

OLJXRTRRJSMURJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao-Cheng Chen et al.
International journal of molecular sciences, 21(18) (2020-09-13)
Malignant melanoma is the most harmful type of skin cancer and its incidence has increased in this past decade. Early diagnosis and treatment are urgently desired. In this study, we conjugated picolinamide/nicotinamide with the pharmacophore of 131I-MIP-1145 to develop 131I-iodofluoropicolinamide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service