Skip to Content
Merck
All Photos(1)

Key Documents

641790

Sigma-Aldrich

Violanthrone-79

Synonym(s):

16,17-Bis(octyloxy)anthra[9,1,2-cde-]benzo[rst]pentaphene-5,10-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H48O4
CAS Number:
Molecular Weight:
712.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

193-196 °C (lit.)

λmax

633 nm in toluene

SMILES string

CCCCCCCCOc1cc2-c3ccccc3C(=O)c4ccc5c6ccc7C(=O)c8ccccc8-c9cc(OCCCCCCCC)c(c1c5c24)c6c79

Application

An n-channel organic semiconductor.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Newman, C.R., et al.
Chemistry of Materials, 16, 4436-4436 (2004)
Jenée D Cyran et al.
The Journal of chemical physics, 142(21), 212435-212435 (2015-06-08)
Two-dimensional infrared (2D IR) spectroscopy was used to characterize the structure of a self-assembled polycyclic aromatic hydrocarbon (PAH), violanthrone-79. A local mode basis was constructed using spectroscopic and computational results of anthrone and monomer violanthrone-79. The vibrational modes in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service