637858
2-Keto-3-(methyl-d3)-butyric acid-1,2,3,4-13C4, 3-d sodium salt
99 atom % 13C, 98 atom % D, ≥99% (CP)
Synonym(s):
α-Keto-3-(methyl-d3)-butyric acid-1,2,3,4-13C4,3-d sodium, α-Ketoisovaleric acid-13C4,d3 sodium salt, 2-Ketoisovaleric acid-13C4,d3 sodium salt, 3-Methyl-d3--2-oxobutanoic acid-13C4,d sodium salt, Sodium α-ketoisovalerate-3-(methyl-d3),1,2,3,4-13C4,3-d sodium salt
About This Item
Recommended Products
isotopic purity
99 atom % 13C
98 atom % D
Quality Level
Assay
≥99% (CP)
form
solid
technique(s)
bio NMR: suitable
mp
227-230 °C (lit.)
mass shift
M+8
SMILES string
[Na+].[2H]C([2H])([2H])[13C]([2H])([13CH3])[13C](=O)[13C]([O-])=O
InChI
1S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1/i1D3,2+1,3+1D,4+1,5+1;
InChI key
WIQBZDCJCRFGKA-XCPSOYJTSA-M
Related Categories
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service