Skip to Content
Merck
All Photos(1)

Key Documents

607541

Sigma-Aldrich

2-Ketobutyric acid-13C4,3,3-d2 sodium salt hydrate

99 atom % 13C, 98 atom % D, 98% (CP)

Synonym(s):

α-Keto-butyric acid-4-13C4-3,3-d2 sodium, α-Ketobutyric acid-4-13C4-3,3-d2 sodium salt, 2-Oxobutanoic acid-13C4,3,3-d2 sodium salt, Sodium α-ketobutyrate-13C4,3,3-d2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$653.00
500 MG
$1,630.00

$653.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$653.00
500 MG
$1,630.00

About This Item

Linear Formula:
13CH313CD213CO13CO2Na · xH2O
Molecular Weight:
130.05 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$653.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C
98 atom % D

Quality Level

Assay

98% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

210 °C (dec.) (lit.)

mass shift

M+6

SMILES string

O.[Na+].[2H][13C]([2H])([13CH3])[13C](=O)[13C]([O-])=O

InChI

1S/C4H6O3.Na.H2O/c1-2-3(5)4(6)7;;/h2H2,1H3,(H,6,7);;1H2/q;+1;/p-1/i1+1,2+1D2,3+1,4+1;;

InChI key

PVLJVKQSCAHPPR-WVMSAXHDSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soumya P Behera et al.
Nature communications, 11(1), 5547-5547 (2020-11-05)
Methyl-NMR enables atomic-resolution studies of structure and dynamics of large proteins in solution. However, resonance assignment remains challenging. The problem is to combine existing structural informational with sparse distance restraints and search for the most compatible assignment among the permutations.

Articles

We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service