Skip to Content
Merck
All Photos(1)

Key Documents

634573

Sigma-Aldrich

ETT Activator

0.25 M in acetonitrile

Synonym(s):

5-(Ethylthio)-1H-tetrazole, 5-(Ethylthio)tetrazole solution

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 2 L
$3,440.00

$3,440.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
4 X 2 L
$3,440.00

About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

$3,440.00


Check Cart for Availability

Request a Bulk Order

product line

Proligo Reagents

concentration

0.25 M in acetonitrile

impurities

<0.002% water

bp

81-82 °C

density

0.786 g/mL at 25 °C

SMILES string

CCSc1nnn[nH]1

InChI

1S/C3H6N4S/c1-2-8-3-4-6-7-5-3/h2H2,1H3,(H,4,5,6,7)

InChI key

GONFBOIJNUKKST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup - Solvent

Flash Point(C)

6 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service