Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ETT Activator
Synonym(s):
5-(Ethylthio)-1
H
-tetrazole, 5-(Ethylthio)tetrazole solution
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Color
Physical Form
Product Line
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
L030000
ETT Activator,
0.25 M
Expand
View Pricing
L0302511
ETT Activator,
Expand
View Pricing
L030080
ETT Activator,
0.25 M, configured for PerkinElmer, configured for Polygen, configured for MerMade, configured for ABI 3900
Expand
View Pricing
634573
ETT Activator,
0.25 M in acetonitrile
Expand
View Pricing
L030004
ETT Activator,
0.25M
Expand
View Pricing