Skip to Content
Merck
All Photos(1)

Key Documents

630837

Sigma-Aldrich

2-Octyn-1-ol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$76.00

$76.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$76.00

About This Item

Linear Formula:
CH3(CH2)4C≡COH
CAS Number:
Molecular Weight:
126.20
Beilstein:
1744120
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$76.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.4560 (lit.)

bp

76-78 °C/2 mmHg (lit.)

density

0.880 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCC#CCO

InChI

1S/C8H14O/c1-2-3-4-5-6-7-8-9/h9H,2-5,8H2,1H3

InChI key

TTWYFVOMGMBZCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service