Skip to Content
Merck
All Photos(1)

Key Documents

161314

Sigma-Aldrich

3-Butyn-2-one

95%

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 ML
$551.00
10 ML
$1,150.00

$551.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 ML
$551.00
10 ML
$1,150.00

About This Item

Linear Formula:
HC≡CCOCH3
CAS Number:
Molecular Weight:
68.07
Beilstein:
605353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$551.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.406 (lit.)

bp

85 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)C#C

InChI

1S/C4H4O/c1-3-4(2)5/h1H,2H3

InChI key

XRGPFNGLRSIPSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Butyn-2-one undergoes asymmetric double-Michael reaction with ortho-tosylamidophenyl malonate catalyzed by chiral aminophosphines to yield indolines[1]. It undergoes double Michael reaction with nitrogen-containing tethered diacid to give pipecolic acid derivatives[2].

Application

3-Butyn-2-one was used in the synthesis of clerodane diterpenoid (+/-)-sacacarin[3]. It was used as substrate in stereoselective, conjugate arylation mediated by gallium(III) chloride leading to (E)-α,β-unsaturated ketones[4].

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R B Grossman et al.
Organic letters, 3(25), 4027-4030 (2001-12-12)
[reaction: see text] The putative structure of the naturally occurring clerodane diterpenoid (+/-)-sacacarin has been prepared in only 10 steps, six of which are C-C bond-forming steps, in a chemo-, regio-, and diastereoselective manner. The key part of the synthesis
Synlett, 809-809 (2007)
F Hughes et al.
Organic letters, 3(18), 2911-2914 (2001-09-01)
[reaction: see text]. Nitrogen-containing tethered diacids, easily prepared by reductive alkylation of diethyl aminomalonate or ethyl cyanoglycinate, undergo double Michael reactions with 3-butyn-2-one to give highly functionalized and substituted piperidines (pipecolic acid derivatives) with surprisingly high stereoselectivity. The heterocyclic double
San N Khong et al.
Molecules (Basel, Switzerland), 17(5), 5626-5650 (2012-05-15)
The bisphosphine-catalyzed double-Michael addition of dinucleophiles to electron-deficient acetylenes is an efficient process for the synthesis of many nitrogen-containing heterocycles. Because the resulting heterocycles contain at least one stereogenic center, this double-Michael reaction would be even more useful if an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service