Skip to Content
Merck
All Photos(1)

Documents

615587

Sigma-Aldrich

4-Aminobutyric acid-2,2,3,3,4,4-d6

97 atom % D

Synonym(s):

GABA-d6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CD2)3CO2H
CAS Number:
Molecular Weight:
109.16
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

97 atom % D

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

195 °C (dec.) (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)/i1D2,2D2,3D2

InChI key

BTCSSZJGUNDROE-NMFSSPJFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Man-Shan Yau et al.
The Science of the total environment, 673, 327-336 (2019-04-17)
A strategy to construct multivariate biomarkers for exposure to algal neurotoxins via correlating changes to the profiles of a series of neurotransmitters and their metabolites in the central nervous system (CNS) of exposed test organism is reported. 3-Month-old marine medaka
Elva Ngai-Yu Lei et al.
Archives of environmental contamination and toxicology, 72(2), 269-280 (2016-12-19)
The simultaneous profiling of 43 functional metabolites in the brain of the small model vertebrate organism, marine medaka (Oryzais melastigma), has been accomplished via dansyl chloride derivatization and LC-MS/MS quantification. This technique was applied to examine effects of 2,2',4,4'-tetrabromodiphenyl ether
Annelie Abrahamsson et al.
Scientific reports, 7, 42733-42733 (2017-02-16)
Metabolic reprogramming is a hallmark of cancer. Nutrient availability in the tissue microenvironment determines cellular events and may play a role in breast carcinogenesis. High mammographic density is an independent risk factor for breast cancer. Whether nutrient availability differs in
María Encarnación Blanco et al.
Journal of neuroscience methods, 341, 108760-108760 (2020-05-20)
So far, analytical investigation of neuroactive molecules in cerebrospinal fluid (CSF) of rodent models has been limited to rats, given the intrinsic anatomic difficulties related to mice sampling and the corresponding tiny amounts of CSF obtained. This poses a challenge
Zhiying Wang et al.
Communications biology, 3(1), 39-39 (2020-01-24)
Osteoporosis is a highly prevalent chronic aging-related disease that frequently is only detected after fracture. We hypothesized that aminobutyric acids could serve as biomarkers for osteoporosis. We developed a quick, accurate, and sensitive screening method for aminobutyric acid isomers and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service