Skip to Content
Merck
All Photos(1)

Key Documents

616281

Sigma-Aldrich

L-Glutamic acid-2,3,3,4,4-d5

97 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$839.00

$839.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$839.00

About This Item

Linear Formula:
HO2C(CD2)2CD(NH2)CO2H
Molecular Weight:
152.16
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$839.00


Check Cart for Availability

Request more information

isotopic purity

97 atom % D

Assay

98% (CP)

form

solid

optical activity

[α]25/D +31°, c = 2 in 5 M HCl

mp

205 °C (dec.) (lit.)

mass shift

M+5

SMILES string

[2H]C([2H])(C(O)=O)C([2H])([2H])[C@]([2H])(N)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1/i1D2,2D2,3D

InChI key

WHUUTDBJXJRKMK-NKXUJHECSA-N

Related Categories

Biochem/physiol Actions

Glutamic acid (or glutamate) functions as a neurotransmitter and also serves as a precursor to other neurotransmitters such as gamma-aminobutyric acid.[1] Glutamic acid also plays a key role in many metabolic pathways that controls growth, reproduction, maintenance and immunity.[2] It is converted to α-ketoglutarate, a key component of the TCA (tricarboxylic acid) cycle and a precursor for the biosynthesis of nucleic acids and certain amino acids.[2][3] In cells, glutamine is converted to glutamate by the enzyme glutaminase. [3]
Appl- L-Glutamic acid-2,3,3,4,4-d5 has been used as an internal standard in metabolomics and lipidomics profiling.[4]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The biology of cancer: metabolic reprogramming fuels cell growth and proliferation.
DeBerardinis RJ
Cell Metabolism, 7(1), 11-20 (2008)
Flavia Bianchi et al.
Insects, 11(11) (2020-11-22)
The production of phagostimulant and attractive volatile organic compounds (VOCs) by yeasts can be exploited to improve the efficacy of attract-and-kill formulations against the spotted wing drosophila (SWD). This study evaluated the persistence over one week of a yeast-based formulation
Glutamic Acid, Twenty Years Later
S. Garattini
The Journal of Nutrition, 130(4), 901S-909S (2000)
Metabolomics and lipidomics using traveling-wave ion mobility mass spectrometry.
Paglia G and Astarita G
Nature Protocols, 12(4), 797-797 (2017)
Amino acids: metabolism, functions, and nutrition.
Wu G
Amino Acids, 37(1), 1-17 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service