Skip to Content
Merck
All Photos(1)

Key Documents

601586

Sigma-Aldrich

Boric acid-10B

95 atom % 10B

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$274.00
25 G
$933.00

$274.00


Available to ship on22 April 2025Details


Request more information

Select a Size

Change View
5 G
$274.00
25 G
$933.00

About This Item

Linear Formula:
H310BO3
CAS Number:
Molecular Weight:
61.03
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

$274.00


Available to ship on22 April 2025Details


Request more information

isotopic purity

95 atom % 10B

Quality Level

density

1.435 g/mL at 25 °C (lit.)

mass shift

M-1

SMILES string

O[10B](O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H/i1-1

InChI key

KGBXLFKZBHKPEV-BJUDXGSMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service