Skip to Content
Merck
All Photos(1)

Key Documents

143219

Sigma-Aldrich

4-Bromophenylhydrazine hydrochloride

99%

Synonym(s):

1-(4-Bromophenyl)hydrazine hydrochloride, 2-(4-Bromophenyl)hydrazinium chloride, 4-Bromophenylhydrazine monohydrochloride, p-Bromophenylhydrazine monohydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$299.00
50 G
$1,130.00

$299.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$299.00
50 G
$1,130.00

About This Item

Linear Formula:
BrC6H4NHNH2·HCl
CAS Number:
Molecular Weight:
223.50
Beilstein:
3565838
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$299.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

mp

220-230 °C (dec.) (lit.)

functional group

bromo
hydrazine

SMILES string

Cl.NNc1ccc(Br)cc1

InChI

1S/C6H7BrN2.ClH/c7-5-1-3-6(9-8)4-2-5;/h1-4,9H,8H2;1H

InChI key

RGGOWBBBHWTTRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromophenylhydrazine hydrochloride was used as reagent in the synthesis of:
  • acylsulfonamides and acylsulfamides[1]
  • 9-bromo-2-ethenyl-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Xie et al.
European journal of medicinal chemistry, 40(7), 655-661 (2005-06-07)
The introduction of side chains bearing epoxide motifs into the molecular scaffold of kenpaullone and 9-trifluoromethylpaullone led to improved antiproliferative activity of the novel derivatives for human tumor cell lines. The syntheses were accomplished applying Stille coupling for the introduction
Michael P Winters et al.
Bioorganic & medicinal chemistry letters, 18(6), 1926-1930 (2008-03-01)
A series of novel acylsulfonamide, acylsulfamide, and sulfonylurea bioisosteres of carboxylic acids were prepared as CXCR2 antagonists. Structure-activity relationships are reported for these series. One potent orally bioavailable inhibitor had excellent PK properties and was active in a lung injury
Siva Doddi et al.
Journal of fluorescence, 26(6), 1939-1949 (2016-11-01)
We report the synthesis and characterization of a new fluorescent dyad SP-DPP-SP(9) via efficient palladium-catalyzed Sonogashira coupling of prop-2-yn-1-yl 3-(3',3'dimethyl-6-nitrospiro[chromene-2,2'-indolin]-1'-yl)propanoatespiropyran, SP(8), a well known photochromic accepter, with 3,6-bis(5-bromothiophen-2-yl)-2,5-bis((R)-2-ethylhexyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione, DPP(4), a highly fluorescent donor. Under visible light exposure the SP unit
Ilkoo Noh et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(3), 1700481-1700481 (2018-03-30)
A noninvasive and selective therapy, photodynamic therapy (PDT) is widely researched in clinical fields; however, the lower efficiency of PDT can induce unexpected side effects. Mitochondria are extensively researched as target sites to maximize PDT effects because they play crucial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service