Skip to Content
Merck
All Photos(1)

Key Documents

593990

Sigma-Aldrich

Ammonium-15N2,d8 sulfate

99 atom % 15N, 98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,580.00

$1,580.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$1,580.00

About This Item

Linear Formula:
(15ND4)2SO4
Molecular Weight:
142.18
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

$1,580.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 15N
98 atom % D

form

solid

technique(s)

protein expression: suitable

mp

>280 °C (dec.) (lit.)

mass shift

M+10

SMILES string

[2H][15N+]([2H])([2H])[2H].[2H][15N+]([2H])([2H])[2H].[O-]S([O-])(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)/i2*1+1;/hD8

InChI key

BFNBIHQBYMNNAN-HESUPHTJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service