Skip to Content
Merck
All Photos(1)

Documents

348473

Sigma-Aldrich

Ammonium-15N2 sulfate

10 atom % 15N

Synonym(s):

15N Labeled ammonium sulfate, Diazanium-15N2 sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(15NH4)2SO4
CAS Number:
Molecular Weight:
134.13
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

10 atom % 15N

mol wt

132.34 by atom % calculation (10 atom % 15N)

mp

>280 °C (dec.) (lit.)

application(s)

agriculture

mass shift

M+2

SMILES string

[15NH3].[15NH3].OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)/i2*1+1;

InChI key

BFNBIHQBYMNNAN-ISOLYIDJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium-15N2 sulfate enriched with 15N isotope

Application

Ammonium-15N2 sulfate finds its application
  • In the field of agriculture research.
  • As a marker to identify the digestibility sites in animal cattle.
  • In metabolic labeling and quantification of proteome dynamic studie.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lays Débora Silva Mariz et al.
Journal of animal science, 96(6), 2453-2467 (2018-04-19)
The objectives of this study were to evaluate the effect of reducing dietary CP contents on 1) total and partial nutrient digestion and nitrogen balance and 2) on microbial crude protein (MCP) synthesis and true MCP digestibility in the small
Annette Dreisbach et al.
Proteomics, 8(10), 2062-2076 (2008-05-21)
Bacillus subtilis has been developed as a model system for physiological proteomics. However, thus far these studies have mainly been limited to cytoplasmic, extracellular, and cell-wall attached proteins. Although being certainly important for cell physiology, the membrane protein fraction has

Related Content

Tracer techniques using isotopic N have been employed since the 1940s to investigate such topics as N transformations and cycling in agricultural and other soils, N sources for crop uptake, the fate and behavior of fertilizer N applied to soils, the nature and extent of soil and fertilizer losses, and fertilizer N uptake efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service