Skip to Content
Merck
All Photos(1)

Key Documents

593028

Sigma-Aldrich

trans-4-Octene

98%

Synonym(s):

δ-trans-Octene, (4E)-4-Octene, (E)-4-Octene, trans-1,2-Dipropylethene, trans-1,2-Dipropylethylene, trans-n-4-Octene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$206.00
25 G
$761.00

$206.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$206.00
25 G
$761.00

About This Item

Linear Formula:
CH3CH2CH2CH=CHCH2CH2CH3
CAS Number:
Molecular Weight:
112.21
Beilstein:
1719104
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$206.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.412 (lit.)

bp

122-123 °C (lit.)

density

0.715 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CCC)=C(\[H])CCC

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h7-8H,3-6H2,1-2H3/b8-7+

InChI key

IRUCBBFNLDIMIK-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service