570745
L-Asparagine-15N2,d8 monodeuterate
98 atom % D, 98 atom % 15N
About This Item
Recommended Products
isotopic purity
98 atom % 15N
98 atom % D
form
solid
optical activity
[α]20/D +31.5°, c = 1 in 1 M HCl
technique(s)
bio NMR: suitable
mp
233-235 °C (lit.)
mass shift
M+10
SMILES string
[2H]OC([C@]([15N]([2H])[2H])([2H])C([2H])([2H])C([15N]([2H])[2H])=O)=O
InChI
1S/C4H8N2O3/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H2,6,7)(H,8,9)/t2-/m0/s1/i1D2,2D,5+1,6+1/hD5
InChI key
DCXYFEDJOCDNAF-VYGMAKJPSA-N
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service