Skip to Content
Merck
All Photos(2)

Key Documents

613428

Sigma-Aldrich

Deuterium oxide

70 atom % D

Synonym(s):

Heavy water, Water-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.12

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

70 atom % D

Quality Level

Assay

≥69.5% (ATOM % D, qNMR)

form

liquid

technique(s)

bio NMR: suitable

bp

101.4 °C (lit.)

mp

3.8 °C (lit.)

application(s)

electronics

storage temp.

room temp

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Matthew S Brook et al.
The Journal of physiology, 594(24), 7399-7417 (2016-09-23)
Resistance exercise training (RET) is one of the most effective strategies for preventing declines in skeletal muscle mass and strength with age. Hypertrophic responses to RET with age are diminished compared to younger individuals. In response to 6 weeks RET, we
M V Franchi et al.
Acta physiologica (Oxford, England), 223(3), e13056-e13056 (2018-02-14)
We assessed focal adhesion kinase (FAK) response to concentric (CON) vs eccentric (ECC) resistance training (RT) at two vastus lateralis (VL) sites, and the relationships between FAK, muscle protein synthesis (MPS) and morphological remodelling. Six young males trained both legs
Nicholas J Saner et al.
The Journal of physiology, 598(8), 1523-1536 (2020-02-23)
Sleep restriction has previously been associated with the loss of muscle mass in both human and animal models. The rate of myofibrillar protein synthesis (MyoPS) is a key variable in regulating skeletal muscle mass and can be increased by performing
Casper van der Wel et al.
Biophysical journal, 113(5), 1037-1046 (2017-09-07)
Understanding interactions between microparticles and lipid membranes is of increasing importance, especially for unraveling the influence of microplastics on our health and environment. Here, we study how a short-ranged adhesive force between microparticles and model lipid membranes causes membrane-mediated particle
Sandra J Veen et al.
Physical review letters, 109(24), 248302-248302 (2013-02-02)
By using the critical Casimir force, we study the attractive strength dependent aggregation of colloids with and without gravity by means of near field scattering. Significant differences were seen between microgravity and ground experiments, both in the structure of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service