Skip to Content
Merck
All Photos(1)

Key Documents

554243

Sigma-Aldrich

3-Chloro-4-methylbenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)SO2Cl
CAS Number:
Molecular Weight:
225.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

34-38 °C (lit.)

SMILES string

Cc1ccc(cc1Cl)S(Cl)(=O)=O

InChI

1S/C7H6Cl2O2S/c1-5-2-3-6(4-7(5)8)12(9,10)11/h2-4H,1H3

InChI key

GNYVVCRRZRVBDD-UHFFFAOYSA-N

General description

3-Chloro-4-methylbenzenesulfonyl chloride can be prepared from 4-methylbenzenesulfonyl chloride via chlorination.[1]

Application

3-Chloro-4-methylbenzenesulfonyl chloride may be used in the synthesis of 2-chloro 4-methylsulfonamide benzylamine.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven J Taylor et al.
Bioorganic & medicinal chemistry letters, 19(20), 5864-5868 (2009-09-18)
A series of potent nicotinamide inhibitors of soluble epoxides hydrolase (sEH) is disclosed. This series was designed using structure-based deconstruction and a combination of two HTS hit series, resulting in hybrid analogs that retained the optimal potency from one series
Wiley-VCH
Ullmann's Fine Chemicals, 295-295 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service