Skip to Content
Merck
All Photos(1)

Key Documents

552429

Sigma-Aldrich

Ethyl 4-hydroxy-2-methyl-1,1-dioxo-2H-1,2-benzothiazine-3-carboxylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$550.00

$550.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$550.00

About This Item

Empirical Formula (Hill Notation):
C12H13NO5S
CAS Number:
Molecular Weight:
283.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$550.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

140-144 °C (lit.)

functional group

ester
ketone
sulfonamide

SMILES string

CCOC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1C

InChI

1S/C12H13NO5S/c1-3-18-12(15)10-11(14)8-6-4-5-7-9(8)19(16,17)13(10)2/h4-7,14H,3H2,1-2H3

InChI key

QWTNANUGXZEPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service