Skip to Content
Merck
All Photos(1)

Key Documents

549231

Sigma-Aldrich

3-Ethoxythiophenol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$184.00
5 G
$509.00

$184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$184.00
5 G
$509.00

About This Item

Linear Formula:
C2H5OC6H4SH
CAS Number:
Molecular Weight:
154.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$184.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.5706 (lit.)

bp

238-239 °C (lit.)

density

1.096 g/mL at 25 °C (lit.)

SMILES string

CCOc1cccc(S)c1

InChI

1S/C8H10OS/c1-2-9-7-4-3-5-8(10)6-7/h3-6,10H,2H2,1H3

InChI key

RTJMJGJLVSKSEB-UHFFFAOYSA-N

General description

3-ethoxythiophenol (ETP) ) is a thiol derivative.

Application

3-ethoxythiophenol (ETP) ) may be used to functionalize gold nanoparticles for developing:
  • Partially selective gold nanoparticle chemiresistor sensors for the quantification of benzene, toluene, ethylbenzene, p-xylene and naphthalene (BTEXN) in water samples[1]
  • Flexible and tunable sensor based on monolayer-capped nanoparticles (MCNPs), with potential application in e-skin devices.[2]
  • gold nanoparticle-based flexible sensor array[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tunable touch sensor and combined sensing platform: Toward nanoparticle-based electronic skin.
Segev-Bar M, et al.
ACS Applied Materials & Interfaces, 5(12), 5531-5541 (2013)
Self-Healable Sensors Based Nanoparticles for Detecting Physiological Markers via Skin and Breath: Toward Disease Prevention via Wearable Devices.
Jin H, et al.
Nano Letters, 16(7), 4194-4202 (2016)
J S Cooper et al.
The Analyst, 140(9), 3233-3238 (2015-03-15)
Partially selective gold nanoparticle sensors have the sensitivity and selectivity to discriminate and quantify benzene, toluene, ethylbenzene, p-xylene and naphthalene (BTEXN) at concentrations relevant to the US Environmental Protection Agency. In this paper we demonstrate that gold nanoparticle chemiresistors can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service