Skip to Content
Merck
All Photos(2)

Key Documents

545910

Sigma-Aldrich

3-(4-Fluorophenoxy)propionic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4O(CH2)2CO2H
CAS Number:
Molecular Weight:
184.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

83-85 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)CCOc1ccc(F)cc1

InChI

1S/C9H9FO3/c10-7-1-3-8(4-2-7)13-6-5-9(11)12/h1-4H,5-6H2,(H,11,12)

InChI key

QCEDXODAYRWQRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(4-Fluorophenoxy)propionic acid can be synthesized by employing 3-(4-fluorophenoxy)propanenitrile as a starting reagent.

Application

3-(4-Fluorophenoxy)propionic acid may be used as one of the reactants for the synthesis of 2,3-dihydro-6-fluoro-4H-1-benzopyran-4-one.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, absolute configuration and conformation of the aldose reductase inhibitor sorbinil
Sarges R, et al.
Journal of Medicinal Chemistry, 28.11, 1716-1720 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service