Skip to Content
Merck
All Photos(2)

Key Documents

546569

Sigma-Aldrich

1,1-Bis(hydroxymethyl)cyclopropane

90%

Synonym(s):

Cyclopropanedimethanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$92.50

$92.50


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
$92.50

About This Item

Linear Formula:
(HOCH2)2C3H4
CAS Number:
Molecular Weight:
102.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.50


Estimated to ship on27 April 2025


Request a Bulk Order

Quality Level

Assay

90%

refractive index

n20/D 1.4700 (lit.)

bp

235-236 °C (lit.)

density

1.065 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC1(CO)CC1

InChI

1S/C5H10O2/c6-3-5(4-7)1-2-5/h6-7H,1-4H2

InChI key

YAINYZJQSQEGND-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service