Skip to Content
Merck
All Photos(1)

Key Documents

544388

Sigma-Aldrich

4-(1-Hydroxyethyl)aniline

97%

Synonym(s):

1-(4-Aminophenyl)ethanol, 4-Amino-α-methylbenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH(CH3)OH
CAS Number:
Molecular Weight:
137.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

70-74 °C (lit.)

SMILES string

CC(O)c1ccc(N)cc1

InChI

1S/C8H11NO/c1-6(10)7-2-4-8(9)5-3-7/h2-6,10H,9H2,1H3

InChI key

JDEYBJHOTWGYFE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behrouz Mohammadi Nargesi et al.
Frontiers in bioengineering and biotechnology, 6, 201-201 (2019-01-22)
Aromatic amines are an important class of chemicals which are used as building blocks for the synthesis of polymers and pharmaceuticals. In this study we establish a de novo pathway for the biosynthesis of the aromatic amines para-amino-phenylethanol (PAPE) and
Study of liquid-phase dehydration of d,l-1-(4-aminophenyl) ethanol in the presence of acid catalysts.
Schul?tsev AL and Panarin EF.
Russ. J. Gen. Chem., 80(7), 1309-1313 (2010)
Birhanu Desalegn Assresahegn et al.
ChemSusChem, 10(20), 4080-4089 (2017-08-24)
In this work, the effects of the formulation of silicon-based composite anodes on their mechanical, storage, and electrochemical properties were investigated. The electrode formulation was changed through the use of hydrogenated or modified (through the covalent attachment of a binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service