Skip to Content
Merck
All Photos(1)

Key Documents

544221

Sigma-Aldrich

2-Fluoro-5-nitrobenzonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$71.20

$71.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$71.20

About This Item

Linear Formula:
FC6H3(CN)NO2
CAS Number:
Molecular Weight:
166.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$71.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

76-80 °C (lit.)

functional group

fluoro
nitrile
nitro

SMILES string

[O-][N+](=O)c1ccc(F)c(c1)C#N

InChI

1S/C7H3FN2O2/c8-7-2-1-6(10(11)12)3-5(7)4-9/h1-3H

InChI key

YLACBMHBZVYOAP-UHFFFAOYSA-N

General description

2-Fluoro-5-nitrobenzonitrile can be prepared from 2-fluorobenzonitrile via nitration.[1]

Application

2-Fluoro-5-nitrobenzonitrile may be used in the preparation of:
  • 2-fluoro-5-aminobenzonitrile[1]
  • ethyl 3-amino-5-nitro-benzo[b]thiophene-2-carboxylate[2]
  • 1-methyl-5-nitro-1H-indazol-3-ylamine[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mimi L Quan et al.
Journal of medicinal chemistry, 48(6), 1729-1744 (2005-03-18)
Modification of a series of pyrazole factor Xa inhibitors to incorporate an aminobenzisoxazole as the P(1) ligand resulted in compounds with improved selectivity for factor Xa relative to trypsin and plasma kallikrein. Further optimization of the P(4) moiety led to
Synthesis, antiproliferative activity, and in silico insights of new 3-benzoylamino-benzo [b] thiophene derivatives.
Martorana A, et al.
European Journal of Medicinal Chemistry, 90, 537-546 (2015)
A method for the regioselective synthesis of 1-alkyl-1H-indazoles.
Liu HJ, et al.
Tetrahedron, 69(19), 3907-3912 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service