Skip to Content
Merck
All Photos(1)

Documents

365831

Sigma-Aldrich

5-Fluoro-2-nitrobenzotrifluoride

99%

Synonym(s):

2-Nitro-α,α,α,5-tetrafluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)CF3
CAS Number:
Molecular Weight:
209.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.46 (lit.)

bp

198-199 °C (lit.)

mp

23 °C (lit.)

density

1.497 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(F)cc1C(F)(F)F

InChI

1S/C7H3F4NO2/c8-4-1-2-6(12(13)14)5(3-4)7(9,10)11/h1-3H

InChI key

WMQOSURXFLBTPC-UHFFFAOYSA-N

General description

5-Fluoro-2-nitrobenzotrifluoride is formed by the nitration of m-fluorobenzotrifluoride.

Application

5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brown JH, et al.
Journal of the Chemical Society, S95-S99 (1949)
Fluorinated high-performance polymers: Poly (arylene ether) s and aromatic polyimides containing trifluoromethyl groups.
Dhara MG and Banerjee S.
Progress in Polymer Science, 35(8), 1022-1027 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service