Skip to Content
Merck
All Photos(1)

Key Documents

542881

Sigma-Aldrich

2-(Trifluoromethoxy)phenylacetonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4CH2CN
CAS Number:
Molecular Weight:
201.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.449 (lit.)

bp

222-223 °C (lit.)

density

1.28 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)Oc1ccccc1CC#N

InChI

1S/C9H6F3NO/c10-9(11,12)14-8-4-2-1-3-7(8)5-6-13/h1-4H,5H2

InChI key

OIKWCWVMUBCXJM-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fused thiophene derivatives as MEK inhibitors
Laing VE, et al.
Bioorganic & Medicinal Chemistry Letters, 22.1, 472-475 (2012)
Selectivity issues in the catalytic multiphase reduction of functionalized halogenated aromatics over Pd/C, Pt/C, and Raney-Ni
Evdokimova, Galina, et al.
Applied Catalysis A: General, 271.1 , 129-136 (2004)
Synthesis of Phloroglucinol Monoaryl Ethers
Sherwood AM, et al.
Synthesis, 44.08, 1208-1212 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service