Skip to Content
Merck
All Photos(2)

Key Documents

516953

Sigma-Aldrich

Mercury(II) nitrate monohydrate

≥99.99% trace metals basis

Synonym(s):

Mercuric nitrate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(NO3)2 · H2O
CAS Number:
Molecular Weight:
342.62
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.99% trace metals basis

reaction suitability

core: mercury
reagent type: catalyst

impurities

<100 ppm total metallic impurities

reduction residue

≤0.01%

mp

79 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤0.001%
Hg+: ≤0.2%

SMILES string

O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Hg.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

KVICROHOONHSRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for oxymercuration; amidomercuration; cyclopropanecleavage; glycosylation

Features and Benefits

Meets A.C.S. reagent specifications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Collum, D. B.; Still, W. C.; Mohamadi, F.
Journal of the American Chemical Society, 105, 6882-6882 (1983)
Kocovsky, P.,
Organometallic News, 12, 1969-1969 (1993)
Wiesner, K.; Tsai, T. Y. R.; Jin, H.
Helvetica Chimica Acta, 68, 300-300 (1985)
Fry, A. J.; Simon, J. A.
The Journal of Organic Chemistry, 47, 5032-5032 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service