Skip to Content
Merck
All Photos(2)

Key Documents

680265

Sigma-Aldrich

Mercury(I) nitrate dihydrate

reagent grade, ≥97%

Synonym(s):

Mercurous nitrate dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$171.00

$171.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
$171.00

About This Item

Linear Formula:
Hg2(NO3)2 · 2H2O
CAS Number:
Molecular Weight:
561.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
powder or crystals

$171.00


Available to ship on17 April 2025Details


Request a Bulk Order

grade

reagent grade

Quality Level

Assay

≥97%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: mercury

density

4.78 g/mL at 25 °C (lit.)

SMILES string

[Hg+].[H]O[H].[O-][N+]([O-])=O

InChI

1S/Hg.NO3.H2O/c;2-1(3)4;/h;;1H2/q+1;-1;

InChI key

UPCSQZXLTBTBCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mercury(I) nitrate dihydrate can be used as a catalyst for the oxidation of aromatic methyl groups to aldehydes and carboxylic acids. It is also used in the preparation of other mercury(I) compounds. Additionally, mercury(I) nitrate dihydrate acts as a reducing agent.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is there a way to determine the age of a chemical based on lot number? We have a bottle of Mercury (I) nitrate dihydrate in our chemical closet, but are not sure when it was purchased since I was not here when it was originally purchased

    1 answer
    1. Yes, the lot/source number on the bottle can be used to pull up the COA on the product page. On the COA it will give a quality release date to determine how old the product is.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service