Skip to Content
Merck
All Photos(1)

Documents

514802

Sigma-Aldrich

Molybdenum

foil, thickness 0.05 mm, ≥99.9% trace metals basis

Synonym(s):

Molybdenum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
EC Number:
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:

Assay

≥99.9% trace metals basis

form

foil

resistivity

5.0 μΩ-cm, 20°C

thickness

0.05 mm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

Quantity

1.2 g = 50 × 50 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear nonproliferation. Iranian 'medical' reactor stokes search for alternatives.
Richard Stone
Science (New York, N.Y.), 340(6139), 1392-1392 (2013-06-22)
Copper ameliorates fluoride toxicity in fluoride and molybdenum fed rabbits.
Arjun L Khandare et al.
Biomedical and environmental sciences : BES, 26(4), 311-313 (2013-03-29)
Lingxing Zeng et al.
ACS applied materials & interfaces, 5(6), 2182-2187 (2013-02-27)
In the present work, the nanocomposite of MoO2-ordered mesoporous carbon (MoO2-OMC) was synthesized for the first time using a carbon thermal reduction route and the mesoporous carbon as the nanoreactor. The synthesized nanocomposite was characterized by X-ray diffraction (XRD), thermogravimetric
Saleh A Mohamed et al.
BMC complementary and alternative medicine, 13, 40-40 (2013-02-26)
Chewing stick (miswak Salvadora persica L.) is an effective tool for oral hygiene. It possessed various biological properties including significant antibacterial and anti-fungal effects. In the present study, we evaluated the antioxidant compounds in miswak. Miswak root was extracted with
K Cory Macleod et al.
Nature chemistry, 5(7), 559-565 (2013-06-22)
The reduction of gaseous nitrogen is a challenge for industrial, biological and synthetic chemists. Major goals include understanding the formation of ammonia for agriculture, and forming N-C and N-Si bonds for the synthesis of fine chemicals. The iron-molybdenum active site

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service