Skip to Content
Merck
All Photos(1)

Key Documents

517860

Sigma-Aldrich

Phenyl-d5-boronic acid

98 atom % D, 98% (CP)

Synonym(s):

(Pentadeuterophenyl)boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$281.00

$281.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$281.00

About This Item

Linear Formula:
C6D5B(OH)2
CAS Number:
Molecular Weight:
126.96
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$281.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

mp

217-220 °C (lit.)

application(s)

electronics
pharmaceutical

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(B(O)O)c([2H])c1[2H]

InChI

1S/C6H7BO2/c8-7(9)6-4-2-1-3-5-6/h1-5,8-9H/i1D,2D,3D,4D,5D

InChI key

HXITXNWTGFUOAU-RALIUCGRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service